missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-thiophenecarboxylic acid, 96%

Catalog No. AC426840050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426840050 5 g Glass bottle
AC426840010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426840050 Supplier Thermo Scientific Chemicals Supplier No. 426840050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24287-95-4 | C5H3BrO2S | 207.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24287-95-4
Molecular Formula C5H3BrO2S
Molecular Weight (g/mol) 207.04
MDL Number MFCD05865213
InChI Key RVSXMPCELBYUSF-UHFFFAOYSA-N
Synonym 2-bromo-3-thiophenecarboxylic acid, 2-bromo-thiophene-3-carboxylic acid, 2-bromo-thiophene-3-carboxylicacid, 3-thiophenecarboxylic acid, 2-bromo, 2-bromo-3-thenoic acid, acmc-209xc0, 2-bromo-3-carboxythiophene, 2-bromothiophene-3-carboxylic?acid, 2-bromo-3-thiophene carboxylic acid, 2-bromo-thiophene 3-carboxylic acid
PubChem CID 12320971
IUPAC Name 2-bromothiophene-3-carboxylic acid
SMILES C1=CSC(=C1C(=O)O)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 179.0°C to 181.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 207.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-thiophenecarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only