missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylthiophene 98.0+%, TCI America™

Catalog No. B10255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10255G 5 g
B102525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10255G Supplier TCI America Supplier No. B10255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14282-76-9
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.059
MDL Number MFCD00059741
InChI Key YYJBWYBULYUKMR-UHFFFAOYSA-N
Synonym thiophene, 2-bromo-3-methyl, 2-bromo-3-methyl-thiophene, 1-bromo-2-methyl thiophene, pubchem5191, 2-bromo-3methylthiophene, 2-bromo-3-methythiophene, acmc-1buh4, 2-bromo-3methyl thiophene, 2-bromo-3-methyl thiophene, ksc491i3r
PubChem CID 84314
IUPAC Name 2-bromo-3-methylthiophene
SMILES CC1=C(SC=C1)Br

Specifications

Color Yellow
Boiling Point 176°C
Quantity 5 g
UN Number 3532
Formula Weight 177.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-methylthiophene
EINECSNumber (5)-6412
TSCA No
Recommended Storage Refrigerator