accessibility menu, dialog, popup

UserName

CAS RN 14282-76-9

IUPAC Name: 2-bromo-3-methylthiophene
Synonyms: 1-bromo-2-methyl thiophene
Molecular Weight (g/mol): 177.06
Molecular Formula: C5H5BrS
InChi Key: YYJBWYBULYUKMR-UHFFFAOYSA-N
SMILES: CC1=C(Br)SC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)