missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylthiophene, 97%

Catalog No. p-7058371
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1139306 5 g
AAL1139314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1139306 Supplier Thermo Scientific Chemicals Supplier No. L1139306
Only null left

CAS: 14282-76-9 | C5H5BrS | 177.059 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14282-76-9
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.059
MDL Number MFCD00059741
InChI Key YYJBWYBULYUKMR-UHFFFAOYSA-N
Synonym thiophene, 2-bromo-3-methyl, 2-bromo-3-methyl-thiophene, 1-bromo-2-methyl thiophene, pubchem5191, 2-bromo-3methylthiophene, 2-bromo-3-methythiophene, acmc-1buh4, 2-bromo-3methyl thiophene, 2-bromo-3-methyl thiophene, ksc491i3r
PubChem CID 84314
IUPAC Name 2-bromo-3-methylthiophene
SMILES CC1=C(SC=C1)Br

Specifications

Density 1.58
Boiling Point 173°C to 176°C
Flash Point 68°C (154°F)
Refractive Index 1.571
Quantity 5 g
UN Number UN2810
Beilstein 105719
Formula Weight 177.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-methylthiophene
Hazard Category H227-H302-H318
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 238-175-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only