missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-methylbutyric Acid 90.0+%, TCI America™
$354.17 - $354.17
Chemical Identifiers
| CAS | 565-74-2 |
|---|---|
| Molecular Formula | C5H9BrO2 |
| Molecular Weight (g/mol) | 181.029 |
| MDL Number | MFCD00004210 |
| InChI Key | UEBARDWJXBGYEJ-UHFFFAOYSA-N |
| Synonym | 2-bromo-3-methylbutyric acid, 2-bromoisovaleric acid, butanoic acid, 2-bromo-3-methyl, alpha-bromoisovaleric acid, butyric acid, 2-bromo-3-methyl, 2-bromo-3-methyl-butanoic acid, .alpha.-bromoisovaleric acid, +/--2-bromo-3-methylbutyric acid, bromisovaleriansaure, acmc-209wpb |
| PubChem CID | 11268 |
| IUPAC Name | 2-bromo-3-methylbutanoic acid |
| SMILES | CC(C)C(C(=O)O)Br |
Chemical Identifiers
| 565-74-2 | |
| 181.029 | |
| UEBARDWJXBGYEJ-UHFFFAOYSA-N | |
| 11268 | |
| CC(C)C(C(=O)O)Br |
| C5H9BrO2 | |
| MFCD00004210 | |
| 2-bromo-3-methylbutyric acid, 2-bromoisovaleric acid, butanoic acid, 2-bromo-3-methyl, alpha-bromoisovaleric acid, butyric acid, 2-bromo-3-methyl, 2-bromo-3-methyl-butanoic acid, .alpha.-bromoisovaleric acid, +/--2-bromo-3-methylbutyric acid, bromisovaleriansaure, acmc-209wpb | |
| 2-bromo-3-methylbutanoic acid |
Specifications
| 565-74-2 | |
| 230°C | |
| MFCD00004210 | |
| 3261 | |
| UEBARDWJXBGYEJ-UHFFFAOYSA-N | |
| 2-bromo-3-methylbutanoic acid | |
| 11268 | |
| ≥90.0% (GC) | |
| 2-Bromo-3-methylbutyric Acid |
| White-Yellow | |
| C5H9BrO2 | |
| 25 g | |
| 2-bromo-3-methylbutyric acid, 2-bromoisovaleric acid, butanoic acid, 2-bromo-3-methyl, alpha-bromoisovaleric acid, butyric acid, 2-bromo-3-methyl, 2-bromo-3-methyl-butanoic acid, .alpha.-bromoisovaleric acid, +/--2-bromo-3-methylbutyric acid, bromisovaleriansaure, acmc-209wpb | |
| CC(C)C(C(=O)O)Br | |
| 181.029 | |
| 181.03 | |
| Crystalline Lumps |
Safety and Handling
RTECSNumber : ES8055000
TSCA : Yes