missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7123063
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B36365G 5 g
B363625G 25 g
2 options

Catalog No. B36365G

Supplier: TCI America B36365G

Only null left

Chemical Identifiers

CAS 53663-39-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00079721
InChI Key LSRTWJCYIWGKCQ-UHFFFAOYSA-N
Synonym 2-bromo-3-methyl-benzoic acid, 2-bromo-m-toluic acid, 2-bromo-3-carboxytoluene, 2-bromo-3-methylbenzoicacid, 3-methyl-2-bromobenzoic acid, benzoic acid, bromo-3-methyl, 2-brom-3-methylbenzoic acid, pubchem3805, acmc-1ay3t, ksc497o4r
PubChem CID 2735588
IUPAC Name 2-bromo-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1Br)C(=O)O

Specifications

Melting Point 137°C
Color Yellow
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-methylbenzoic Acid