missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(methoxymethoxy)pyridine, 96%

Catalog No. AAH5232406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5232406 5 g
AAH5232403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5232406 Supplier Thermo Scientific Chemicals Supplier No. H5232406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 162271-10-5 | C7H8BrNO2 | 218.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162271-10-5
Molecular Formula C7H8BrNO2
Molecular Weight (g/mol) 218.05
MDL Number MFCD17015031
InChI Key CATUEJFGWSUPBV-UHFFFAOYSA-N
Synonym 2-bromo-3-methoxymethoxy pyridine, pyridine, 2-bromo-3-methoxymethoxy, 2-bromo-3-methoxymethoxy-pyridine, 2-bromo-3-methoxymethyloxypyridine
PubChem CID 11586573
IUPAC Name 2-bromo-3-(methoxymethoxy)pyridine
SMILES COCOC1=C(N=CC=C1)Br

Specifications

Melting Point 35°C to 37°C
Quantity 5 g
Formula Weight 218.05
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-(methoxymethoxy)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only