accessibility menu, dialog, popup

UserName

CAS RN 162271-10-5

IUPAC Name: 2-bromo-3-(methoxymethoxy)pyridine
Synonyms: 2-bromo-3-(methoxymethoxy)pyridine
Molecular Weight (g/mol): 218.05
Molecular Formula: C7H8BrNO2
InChi Key: CATUEJFGWSUPBV-UHFFFAOYSA-N
SMILES: COCOC1=C(Br)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results