missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-methoxyacetophenone, 97%

CAS: 5000-65-7 | C9H9BrO2 | 229.07 g/mol

$352.88 - $352.88

Chemical Identifiers

CAS 5000-65-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00000199
InChI Key IOOHBIFQNQQUFI-UHFFFAOYSA-N
Synonym 2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide
PubChem CID 101294
IUPAC Name 2-bromo-1-(3-methoxyphenyl)ethanone
SMILES COC1=CC=CC(=C1)C(=O)CBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151910250
SDS View Documents
Thermo Scientific Chemicals
151910250
25 g Glass bottle
Each for $352.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5000-65-7
229.07
IOOHBIFQNQQUFI-UHFFFAOYSA-N
101294
COC1=CC=CC(=C1)C(=O)CBr
C9H9BrO2
MFCD00000199
2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide
2-bromo-1-(3-methoxyphenyl)ethanone

Specifications

5000-65-7
100.0
Green-Yellow to White
97%
C9H9BrO2
MFCD00000199
2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide
COC1=CC=CC(=C1)C(=O)CBr
229.07
229.07
Crystalline Powder
96.0
59°C to 64°C
Authentic
Glass bottle
CH3OC6H4COCH2Br
25 g
IOOHBIFQNQQUFI-UHFFFAOYSA-N
2-bromo-1-(3-methoxyphenyl)ethanone
101294
97%
2-Bromo-3'-methoxyacetophenone, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 225-666-1

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only