missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-hydroxypyridine, 99%

Catalog No. AAA1268706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1268706 5 g
AAA1268714 25 g
AAA1268722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1268706 Supplier Thermo Scientific Chemicals Supplier No. A1268706

CAS: 6602-32-0 | C5H4BrNO | 174.00 g/mol

2-Bromo-3-hydroxypyridine is used to produce 2-bromo-4,6-diiodo-3-pyridinol. It is also used in azo dyes and in the synthesis of pterocellin A by reacting with kojic acid and in synthesis of orelline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-hydroxypyridine is used to produce 2-bromo-4,6-diiodo-3-pyridinol. It is also used in azo dyes and in the synthesis of pterocellin A by reacting with kojic acid and in synthesis of orelline.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 6602-32-0
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174.00
MDL Number MFCD00006220
InChI Key YKHQFTANTNMYPP-UHFFFAOYSA-N
Synonym 2-bromo-3-hydroxypyridine, 2-bromo-3-pyridinol, 3-pyridinol, 2-bromo, 2-bromo-pyridin-3-ol, 3-hydroxy-2-bromopyridine, unii-9dab85h359, 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo3-pyridinol
PubChem CID 23070
IUPAC Name 2-bromopyridin-3-ol
SMILES OC1=CC=CN=C1Br

Specifications

Melting Point 182°C to 186°C
Quantity 5 g
Beilstein 109829
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 174
Percent Purity 99%
Chemical Name or Material 2-Bromo-3-hydroxypyridine
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 229-547-5
RTECSNumber UU7705000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only