missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-hexyl-5-iodothiophene (stabilized with Copper chip) 97.0+%, TCI America™
$193.86 - $663.60
Chemical Identifiers
| CAS | 160096-76-4 |
|---|---|
| Molecular Formula | C10H14BrIS |
| Molecular Weight (g/mol) | 373.09 |
| MDL Number | MFCD15144651 |
| InChI Key | SDMKEQYHISDGKT-UHFFFAOYSA-N |
| PubChem CID | 11474236 |
| IUPAC Name | 2-bromo-3-hexyl-5-iodothiophene |
| SMILES | CCCCCCC1=C(SC(=C1)I)Br |
Chemical Identifiers
| 160096-76-4 | |
| 373.09 | |
| SDMKEQYHISDGKT-UHFFFAOYSA-N | |
| 2-bromo-3-hexyl-5-iodothiophene |
| C10H14BrIS | |
| MFCD15144651 | |
| 11474236 | |
| CCCCCCC1=C(SC(=C1)I)Br |
Specifications
| 160096-76-4 | |
| 121°C | |
| MFCD15144651 | |
| SDMKEQYHISDGKT-UHFFFAOYSA-N | |
| 2-bromo-3-hexyl-5-iodothiophene | |
| 11474236 | |
| ≥97.0% (GC) | |
| 2-Bromo-3-hexyl-5-iodothiophene (stabilized with Copper chip) |
| Yellow | |
| C10H14BrIS | |
| 1 g | |
| CCCCCCC1=C(SC(=C1)I)Br | |
| 373.09 | |
| 373.09 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Freezer