accessibility menu, dialog, popup

UserName

CAS RN 160096-76-4

H3CSIBr

CAS RN 160096-76-4

IUPAC Name: 2-bromo-3-hexyl-5-iodothiophene
Synonyms: 2-bromo-3-hexyl-5-iodothiophene
Molecular Weight (g/mol): 373.09
Molecular Formula: C10H14BrIS
InChi Key: SDMKEQYHISDGKT-UHFFFAOYSA-N
SMILES: CCCCCCC1=C(Br)SC(I)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results