missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals

Catalog No. p-7041341
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6178514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6178514 Supplier Thermo Scientific Chemicals Supplier No. H6178514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59907-13-0 | Molecular Formula: C7H6BrF | Molecular Weight: 189.027 g/mol

2-Bromo-3-fluorotoluene is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluorotoluene is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 59907-13-0
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD08458010
InChI Key FYCXRRYRNRDSRM-UHFFFAOYSA-N
Synonym 2-bromo-3-fluorotoluene, 3-fluoro-2-bromo toluene, benzene, 2-bromo-1-fluoro-3-methyl, 2-brom-1-fluor-3-methylbenzol, pubchem4518, acmc-1awko, ksc493o4f, 2-bromo-1-fluoro-3-methyl-benzene
PubChem CID 108856
IUPAC Name 2-bromo-1-fluoro-3-methylbenzene
SMILES CC1=C(C(=CC=C1)F)Br

Specifications

Melting Point 118°C to 123°C
Density 1.503
Flash Point 76°C (169°F)
Refractive Index 1.533
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 189.02
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-fluorotoluene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 261-981-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only