missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. b33881g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33881G Supplier TCI America Supplier No. B33881G
Only null left

Chemical Identifiers

CAS 104540-42-3
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD00061243
InChI Key UERAGXKMOXUWPC-UHFFFAOYSA-N
Synonym 2-bromo-3-fluorobenzotrifluoride, 2-bromo-1-fluoro-3-trifluoromethyl benzene, 2-fluoro-6-trifluoromethyl bromobenzene, 2-bromo-3-fluoro-1-trifluoromethyl benzene, benzene, 2-bromo-1-fluoro-3-trifluoromethyl, 2-bromo-1-fluoro-3-trifluoromethyl-benzene, 2-bromo-3-fluorobenzo trifluoride, 2-bromo-3-fluorbenzotrifluoride, 2-fluoro-6-trifluoromethyl-bromobenzene
PubChem CID 517847
IUPAC Name 2-bromo-1-fluoro-3-(trifluoromethyl)benzene
SMILES C1=CC(=C(C(=C1)F)Br)C(F)(F)F

Specifications

Color Yellow
Boiling Point 168°C
Quantity 1 g
Formula Weight 243.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-fluorobenzotrifluoride