missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluorobenzoic Acid 96.0+%, TCI America™

Catalog No. B37221G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37221G 1 g
B37225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37221G Supplier TCI America Supplier No. B37221G

Chemical Identifiers

CAS 132715-69-6
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD01569398
InChI Key KQRCBMPPEPNNDS-UHFFFAOYSA-N
Synonym 3-fluoro-2-bromobenzoic acid, 2-bromo-3-fluorobenzoicacid, 2-bromo-3-fluoro-benzoic acid, buttpark 22\01-97, benzoic acid, 2-bromo-3-fluoro, pubchem1319, 2-bromo-3-fluorobenzoic, acmc-1c0re, intermediates-zcf02016, ksc174c5p
PubChem CID 302622
IUPAC Name 2-bromo-3-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)F)Br)C(=O)O

Specifications

Melting Point 160°C
Quantity 1 g
Formula Weight 219.01
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-fluorobenzoic Acid