Learn More
2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
$496.05 - $496.05
Chemical Identifiers
| CAS | 89402-29-9 |
|---|---|
| Molecular Formula | C6H2BrF4N |
| Molecular Weight (g/mol) | 243.987 |
| MDL Number | MFCD10699118 |
| InChI Key | DNVIECHQUYQOOH-UHFFFAOYSA-N |
| PubChem CID | 15592823 |
| IUPAC Name | 2-bromo-3-fluoro-5-(trifluoromethyl)pyridine |
| SMILES | C1=C(C=NC(=C1F)Br)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3199303
|
Thermo Scientific Chemicals
H3199303 |
1 g |
Each for $496.05
|
|
|||||
Description
2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 89402-29-9 | |
| 243.987 | |
| DNVIECHQUYQOOH-UHFFFAOYSA-N | |
| 2-bromo-3-fluoro-5-(trifluoromethyl)pyridine |
| C6H2BrF4N | |
| MFCD10699118 | |
| 15592823 | |
| C1=C(C=NC(=C1F)Br)C(F)(F)F |
Specifications
| 89402-29-9 | |
| 1.464 | |
| 1 g | |
| Not miscible or difficult to mix. | |
| C1=C(C=NC(=C1F)Br)C(F)(F)F | |
| 243.987 | |
| 243.99 | |
| 2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine |
| C6H2BrF4N | |
| MFCD10699118 | |
| 2-bromo-3-fluoro-5-trifluoromethyl pyridine, pyridine,2-bromo-3-fluoro-5-trifluoromethyl, pyridine, 2-bromo-3-fluoro-5-trifluoromethyl, acmc-20a5we, 2-bromo-3-fluoro-5-trifluoromethyl-pyridine | |
| DNVIECHQUYQOOH-UHFFFAOYSA-N | |
| 2-bromo-3-fluoro-5-(trifluoromethyl)pyridine | |
| 15592823 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only