accessibility menu, dialog, popup

UserName

CAS RN 89402-29-9

NFBrFFF

CAS RN 89402-29-9

IUPAC Name: 2-bromo-3-fluoro-5-(trifluoromethyl)pyridine
Synonyms: 2-bromo-3-fluoro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 243.99
Molecular Formula: C6H2BrF4N
InChi Key: DNVIECHQUYQOOH-UHFFFAOYSA-N
SMILES: FC1=C(Br)N=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results