missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAH3199303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3199303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3199303 Supplier Thermo Scientific Chemicals Supplier No. H3199303

2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 89402-29-9
Molecular Formula C6H2BrF4N
Molecular Weight (g/mol) 243.987
MDL Number MFCD10699118
InChI Key DNVIECHQUYQOOH-UHFFFAOYSA-N
PubChem CID 15592823
IUPAC Name 2-bromo-3-fluoro-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1F)Br)C(F)(F)F

Specifications

Refractive Index 1.464
Quantity 1 g
Synonym 2-bromo-3-fluoro-5-trifluoromethyl pyridine, pyridine,2-bromo-3-fluoro-5-trifluoromethyl, pyridine, 2-bromo-3-fluoro-5-trifluoromethyl, acmc-20a5we, 2-bromo-3-fluoro-5-trifluoromethyl-pyridine
Solubility Information Not miscible or difficult to mix.
Formula Weight 243.99
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only