missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,5-difluorophenol, 98%

Catalog No. p-7028795
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2620303 1 g
AAH2620306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2620303 Supplier Thermo Scientific Chemicals Supplier No. H2620303

CAS: 325486-43-9 | C6H3BrF2O | 208.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 325486-43-9
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD01861127
InChI Key AGMQCXYWIMAOMT-UHFFFAOYSA-N
Synonym acmc-20a5l1, phenol,2-bromo-3,5-difluoro, phenol, 2-bromo-3,5-difluoro, 2-bromanyl-3,5-bis fluoranyl phenol
PubChem CID 22144860
IUPAC Name 2-bromo-3,5-difluorophenol
SMILES C1=C(C=C(C(=C1F)Br)O)F

Specifications

Refractive Index 1.529
Quantity 1 g
Formula Weight 208.99
Percent Purity 98%
Chemical Name or Material 2-Bromo-3,5-difluorophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only