Learn More
2-Bromo-3',4'-dichloroacetophenone, 98%
CAS: 2632-10-2 | C8H5BrCl2O | 267.93 g/mol
Supplier: Thermo Scientific Chemicals A1336522
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Bromo-3',4'-dichloroacetophenone | |
| 55°C to 58°C | |
| MFCD00051581 | |
| UN1759 | |
| 2-bromo-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl bromide, 2-bromo-3',4'-dichloroacetophenone, 2-bromo-1-3,4-dichlorophenyl ethan-1-one, 3,4-dichlorophenacylbromide, ethanone, 2-bromo-1-3,4-dichlorophenyl, 1-3,4-dichlorophenyl-2-bromoethan-1-one, pubchem3262, acmc-1coei, 3.4-dichlorophenacylbromide | |
| C1=CC(=C(C=C1C(=O)CBr)Cl)Cl | |
| 267.93 | |
| 267.94 |
| 2632-10-2 | |
| C8H5BrCl2O | |
| 100 g | |
| 511981 | |
| PAKFHEFMTRCFAU-UHFFFAOYSA-N | |
| 2-bromo-1-(3,4-dichlorophenyl)ethanone | |
| 244751 | |
| 98% |
Chemical Identifiers
| 2632-10-2 | |
| 267.93 | |
| PAKFHEFMTRCFAU-UHFFFAOYSA-N | |
| 244751 | |
| C1=CC(=C(C=C1C(=O)CBr)Cl)Cl |
| C8H5BrCl2O | |
| MFCD00051581 | |
| 2-bromo-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl bromide, 2-bromo-3',4'-dichloroacetophenone, 2-bromo-1-3,4-dichlorophenyl ethan-1-one, 3,4-dichlorophenacylbromide, ethanone, 2-bromo-1-3,4-dichlorophenyl, 1-3,4-dichlorophenyl-2-bromoethan-1-one, pubchem3262, acmc-1coei, 3.4-dichlorophenacylbromide | |
| 2-bromo-1-(3,4-dichlorophenyl)ethanone |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only