missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3',4'-dichloroacetophenone, 98%

Catalog No. p-7021145
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1336506 5 g
AAA1336514 25 g
AAA1336522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1336506 Supplier Thermo Scientific Chemicals Supplier No. A1336506
Only null left

CAS: 2632-10-2 | C8H5BrCl2O | 267.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2632-10-2
Molecular Formula C8H5BrCl2O
Molecular Weight (g/mol) 267.93
MDL Number MFCD00051581
InChI Key PAKFHEFMTRCFAU-UHFFFAOYSA-N
Synonym 2-bromo-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl bromide, 2-bromo-3',4'-dichloroacetophenone, 2-bromo-1-3,4-dichlorophenyl ethan-1-one, 3,4-dichlorophenacylbromide, ethanone, 2-bromo-1-3,4-dichlorophenyl, 1-3,4-dichlorophenyl-2-bromoethan-1-one, pubchem3262, acmc-1coei, 3.4-dichlorophenacylbromide
PubChem CID 244751
IUPAC Name 2-bromo-1-(3,4-dichlorophenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CBr)Cl)Cl

Specifications

Melting Point 55°C to 58°C
Quantity 5 g
UN Number UN1759
Beilstein 511981
Formula Weight 267.94
Percent Purity 98%
Chemical Name or Material 2-Bromo-3',4'-dichloroacetophenone
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only