missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. p-7123142
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37491G Supplier TCI America Supplier No. B37491G
Only null left

Chemical Identifiers

CAS 452972-12-2
Molecular Formula C11H15BBrNO2
Molecular Weight (g/mol) 283.96
MDL Number MFCD06798246
InChI Key QAQDSIBZRCSYAR-UHFFFAOYSA-N
Synonym 2-(2-Bromo-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Bromo-3-pyridylboronic Acid Pinacol Ester
PubChem CID 12067061
IUPAC Name 2-bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)Br

Specifications

Melting Point 28°C
Color White-Yellow
Quantity 1 g
Formula Weight 283.96
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine