accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNBr

CAS RN 452972-12-2

IUPAC Name: 2-bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 283.96
Molecular Formula: C11H15BBrNO2
InChi Key: QAQDSIBZRCSYAR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(Br)N=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results