missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-nitropropane, 98%

Catalog No. AAA1838506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1838506 5 g
AAA1838514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1838506 Supplier Thermo Scientific Chemicals Supplier No. A1838506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5447-97-2 | C3H6BrNO2 | 167.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5447-97-2
Molecular Formula C3H6BrNO2
Molecular Weight (g/mol) 167.99
MDL Number MFCD00007389
InChI Key OADSZWXMXIWZSQ-UHFFFAOYSA-N
Synonym propane, 2-bromo-2-nitro, 2-bromo-2-nitro-propane, snxhevlqiijip@, acmc-1aqp4, propane,2-bromo-2-nitro, 2-bromo-2-nitropropane, oadszwxmxiwzsq-uhfffaoysa
PubChem CID 79531
IUPAC Name 2-bromo-2-nitropropane
SMILES CC(C)([N+](=O)[O-])Br

Specifications

Density 1.584
Boiling Point 45°C to 46°C (16 mmHg)
Flash Point 61°C (141°F)
Refractive Index 1.464
Quantity 5 g
Beilstein 1746626
Formula Weight 168
Percent Purity 98%
Chemical Name or Material 2-Bromo-2-nitropropane
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 226-668-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only