missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2-nitro-1,3-propanediol, 98.0-100.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor B22945KGBL
Specifications
| 52-51-7 | |
| Amber Glass Bottle | |
| 5 kg | |
| OCC(Br)(CO)[N+]([O-])=O | |
| 199.99 | |
| Reagent |
| 1 | |
| C3H6BrNO4 | |
| LVDKZNITIUWNER-UHFFFAOYSA-N | |
| 2-bromo-2-nitropropane-1,3-diol | |
| 98.0 to 100.5% | |
| Crystalline Powder or Lumps |
Chemical Identifiers
| 52-51-7 | |
| 199.99 | |
| 2-bromo-2-nitropropane-1,3-diol |
| C3H6BrNO4 | |
| LVDKZNITIUWNER-UHFFFAOYSA-N | |
| OCC(Br)(CO)[N+]([O-])=O |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.