missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropane 98.0+%, TCI America™

Catalog No. p-7118564
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
B061725G 25 g
B0617100G 100 g
B0617500G 500 g
3 options

Catalog No. B061725G

Supplier: TCI America B061725G

Only null left

Chemical Identifiers

CAS 507-19-7
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000125
InChI Key RKSOPLXZQNSWAS-UHFFFAOYSA-N
Synonym tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane
PubChem CID 10485
IUPAC Name 2-bromo-2-methylpropane
SMILES CC(C)(C)Br

Specifications

Color Yellow
Boiling Point 75°C
Quantity 25 g
UN Number 2342
Formula Weight 137.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-methylpropane
EINECSNumber (2)-0074
RTECSNumber TX4150000
TSCA Yes