missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropane 98.0+%, TCI America™

Catalog No. B0617500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0617500G 500 g
B061725G 25 g
B0617100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0617500G Supplier TCI America Supplier No. B0617500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 507-19-7
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000125
InChI Key RKSOPLXZQNSWAS-UHFFFAOYSA-N
Synonym tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane
PubChem CID 10485
IUPAC Name 2-bromo-2-methylpropane
SMILES CC(C)(C)Br

Specifications

Color Yellow
Boiling Point 75°C
Quantity 500 g
UN Number 2342
Formula Weight 137.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-methylpropane
EINECSNumber (2)-0074
RTECSNumber TX4150000
TSCA Yes