missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropane, 96%, stabilized

Catalog No. p-4510368
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159261000 100 mL Glass bottle
AC159265000 500 mL Glass bottle
AC159260025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC159261000 Supplier Thermo Scientific Chemicals Supplier No. 159261000

CAS: 507-19-7 | C4H9Br | 137.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 507-19-7
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000125
InChI Key RKSOPLXZQNSWAS-UHFFFAOYSA-N
Synonym tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane
PubChem CID 10485
IUPAC Name 2-bromo-2-methylpropane
SMILES CC(C)(C)Br

Specifications

Melting Point -20.0°C
Density 1.1800g/mL
Boiling Point 72.0°C to 74.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4269 to 1.4289
Linear Formula (CH3)3CBr
Quantity 100 mL
Beilstein 01,127
Merck Index 15,1557
Specific Gravity 1.18
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Stabilizer Potassium carbonate
Formula Weight 137.02
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-methylpropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 208-065-9

RUO – Research Use Only