missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-methoxyacetophenone, 98%

Catalog No. p-7044674
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0769606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0769606 Supplier Thermo Scientific Chemicals Supplier No. L0769606

CAS: 31949-21-0 | C9H9BrO2 | 229.073 g/mol

2-Bromo-2'-methoxyacetophenone is used as a pharmaceutical intermediate .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2′-methoxyacetophenone is used as a pharmaceutical intermediate .

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 31949-21-0
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00000196
InChI Key GKNCPTLOPRDYMH-UHFFFAOYSA-N
Synonym 2-bromo-1-2-methoxyphenyl ethanone, 2-bromo-2'-methoxyacetophenone, 2-methoxyphenacyl bromide, o-methoxy phenacylbromide, bromomethyl 2-methoxyphenyl ketone, 2-bromo-1-2-methoxyphenyl ethan-1-one, 2-bromo-2'-methoxy acetophenone, ethanone, 2-bromo-1-methoxyphenyl, alpha-bromo-o-methoxyacetophenone, 2-bromoacetylanisole
PubChem CID 123440
IUPAC Name 2-bromo-1-(2-methoxyphenyl)ethanone
SMILES COC1=CC=CC=C1C(=O)CBr

Specifications

Melting Point 42°C to 46°C
Boiling Point 130°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 1938788
Solubility Information Insoluble in water.
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-methoxyacetophenone
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 250-870-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only