accessibility menu, dialog, popup

UserName

CH3OOBr

CAS RN 31949-21-0

IUPAC Name: 2-bromo-1-(2-methoxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(2-methoxyphenyl)ethanone
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: GKNCPTLOPRDYMH-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)CBr
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)