missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-dihydroxyacetophenone 97.0+%, TCI America™

Catalog No. B47671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47671G Supplier TCI America Supplier No. B47671G
Only null left

Chemical Identifiers

CAS 2491-39-6
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD03789098
InChI Key RAULLGKGLGXMOM-UHFFFAOYSA-N
Synonym 2,4-Dihydroxyphenacyl Bromide, 4-(Bromoacetyl)resorcinol
PubChem CID 151017
IUPAC Name 2-bromo-1-(2,4-dihydroxyphenyl)ethanone
SMILES C1=CC(=C(C=C1O)O)C(=O)CBr

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 231.05
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2′,4′-dihydroxyacetophenone