accessibility menu, dialog, popup

UserName

OHHOOBr

CAS RN 2491-39-6

IUPAC Name: 2-bromo-1-(2,4-dihydroxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(2,4-dihydroxyphenyl)ethanone
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: RAULLGKGLGXMOM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C(=O)CBr)C(O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results