missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-13,13-dimethyl-6H-indeno[1,2-b]anthracene-6,11(13H)-dione 98.0+%, TCI America™

Catalog No. b46681g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B46681G Supplier TCI America Supplier No. B46681G

Chemical Identifiers

CAS 1196107-73-9
Molecular Formula C23H15BrO2
Molecular Weight (g/mol) 403.275
MDL Number MFCD20275103
InChI Key NREMKCAHBDRWFB-UHFFFAOYSA-N
PubChem CID 58175361
SMILES CC1(C2=CC3=C(C=C2C4=C1C=C(C=C4)Br)C(=O)C5=CC=CC=C5C3=O)C

Specifications

Melting Point 264°C
Color Yellow
Quantity 1 g
Formula Weight 403.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-13,13-dimethyl-6H-indeno[1,2-b]anthracene-6,11(13H)-dione