accessibility menu, dialog, popup

UserName

CH3BrOOH3C

CAS RN 1196107-73-9

IUPAC Name: 2-bromo-13,13-dimethyl-6H,11H,13H-indeno[1,2-b]anthracene-6,11-dione
Synonyms: 2-bromo-13,13-dimethylindeno[1,2-b]anthracene-6,11-dione
Molecular Weight (g/mol): 403.28
Molecular Formula: C23H15BrO2
InChi Key: NREMKCAHBDRWFB-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC3=C(C=C2C2=C1C=C(Br)C=C2)C(=O)C1=CC=CC=C1C3=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results