missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-phenylpropane 93.0+%, TCI America™

Catalog No. b22905g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22905G Supplier TCI America Supplier No. B22905G

Specifications

CAS 2114-39-8
Molecular Formula C9H11Br
Quantity 5 g
MDL Number MFCD00000149
Synonym 2-bromopropyl benzene, 2-bromo-1-phenylpropane, 1-phenyl-2-bromopropane, 2-bromo-propyl-benzene, benzene, 2-bromopropyl, 2-bromanylpropylbenzene, 2-bromopropyl-benzene, 1-2-bromopropyl benzene, acmc-1cr50
InChI Key NVYOCAOZCSNIHR-UHFFFAOYNA-N
SMILES CC(Br)CC1=CC=CC=C1
IUPAC Name (2-bromopropyl)benzene
Molecular Weight (g/mol) 199.09
PubChem CID 102754
Formula Weight 199.09
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-1-phenylpropane
Show More Show Less