missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-methylbenzimidazole, 97%

Catalog No. AAH60075MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60075MD 250 mg
AAH6007503 1 g
AAH6007506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH60075MD Supplier Thermo Scientific Chemicals Supplier No. H60075MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 49572-60-3 | C8H7BrN2 | 211.06 g/mol

It is a synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a synthetic building block.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 49572-60-3
Molecular Formula C8H7BrN2
Molecular Weight (g/mol) 211.06
MDL Number MFCD00839528
InChI Key PROLKELCEPYEQT-UHFFFAOYSA-N
Synonym 2-bromo-1-methyl-1h-benzo d imidazole, 2-bromo-1-methyl-1h-benzoimidazole, 1h-benzimidazole, 2-bromo-1-methyl, 2-bromo-1-methyl-1h-benzimidazole, 2-bromo-1-methyl-1,3-benzodiazole, 1-methyl-2-bromo-1h-benzimidazole, 2-bromo-1-methyl-1h-1,3-benzodiazole
PubChem CID 797082
SMILES CN1C(Br)=NC2=CC=CC=C12

Specifications

Quantity 250 mg
IUPAC Name 2-bromo-1-methylbenzimidazole
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-methylbenzimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only