CAS RN 49572-60-3
IUPAC Name:
2-bromo-1-methyl-1H-1,3-benzodiazole
Synonyms:
2-bromo-1-methyl-1,3-benzodiazole
Molecular Weight (g/mol):
211.06
Molecular Formula:
C8H7BrN2
InChi Key:
PROLKELCEPYEQT-UHFFFAOYSA-N
SMILES:
CN1C(Br)=NC2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
2-Bromo-1-methylbenzimidazole, 97%
2-Bromo-1-methylbenzimidazole, 97%
CAS: 49572-60-3 Molecular Formula: C8H7BrN2 Molecular Weight (g/mol): 211.06 MDL Number: MFCD00839528 InChI Key: PROLKELCEPYEQT-UHFFFAOYSA-N Synonym: 2-bromo-1-methyl-1h-benzo d imidazole,2-bromo-1-methyl-1h-benzoimidazole,1h-benzimidazole, 2-bromo-1-methyl,2-bromo-1-methyl-1h-benzimidazole,2-bromo-1-methyl-1,3-benzodiazole,1-methyl-2-bromo-1h-benzimidazole,2-bromo-1-methyl-1h-1,3-benzodiazole PubChem CID: 797082 SMILES: CN1C(Br)=NC2=CC=CC=C12