missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-fluoro-4-nitrobenzene 95.0+%, TCI America™

Catalog No. p-7119860
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29585G 5 g
B295825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29585G Supplier TCI America Supplier No. B29585G

Chemical Identifiers

CAS 701-45-1
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 219.997
MDL Number MFCD00055433
InChI Key FAWMTDSAMOCUAR-UHFFFAOYSA-N
Synonym 3-bromo-4-fluoronitrobenzene, 1-bromo-2-fluoro-5-nitrobenzene, 3-bromo-4-fluoro-1-nitrobenzene, 2-bromo-1-fluoro-4-nitro-benzene, 2-bromo-4-nitrofluorobenzene, benzene, 2-bromo-1-fluoro-4-nitro, pubchem8499, acmc-1birt, 3-bromo-4-fluoronitrobezene, ksc494c7t
PubChem CID 69699
IUPAC Name 2-bromo-1-fluoro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Br)F

Specifications

Melting Point 57°C
Quantity 5 g
Formula Weight 220.00
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1-fluoro-4-nitrobenzene