missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-fluoro-4-nitrobenzene, 95%

Catalog No. p-7055459
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6404514 25 g
AAH6404522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6404514 Supplier Thermo Scientific Chemicals Supplier No. H6404514
Only null left

CAS: 701-45-1 | C6H3BrFNO2 | 219.997 g/mol

Reatant in the synthesis of N-fused tricyclic indoles, dimethyamine, benzofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of N-fused tricyclic indoles, dimethyamine, benzofuran.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 701-45-1
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 219.997
MDL Number MFCD00055433
InChI Key FAWMTDSAMOCUAR-UHFFFAOYSA-N
Synonym 3-bromo-4-fluoronitrobenzene, 1-bromo-2-fluoro-5-nitrobenzene, 3-bromo-4-fluoro-1-nitrobenzene, 2-bromo-1-fluoro-4-nitro-benzene, 2-bromo-4-nitrofluorobenzene, benzene, 2-bromo-1-fluoro-4-nitro, pubchem8499, acmc-1birt, 3-bromo-4-fluoronitrobezene, ksc494c7t
PubChem CID 69699
IUPAC Name 2-bromo-1-fluoro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Br)F

Specifications

Quantity 25 g
Beilstein 2089367
Solubility Information Slightly soluble in water.
Formula Weight 220
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromo-1-fluoro-4-nitrobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-855-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only