missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-chloro-4-iodobenzene, 98%

Catalog No. AC458380010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458380010 1 g
AC458380050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458380010 Supplier Thermo Scientific Chemicals Supplier No. 458380010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 31928-46-8 | C6H3BrClI | 317.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31928-46-8
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.35
MDL Number MFCD07780667
InChI Key KSCPVCBJIZDZER-UHFFFAOYSA-N
Synonym 1-bromo-2-chloro-5-iodobenzene, 2-bromo-1-chloro-4-iodo-benzene, benzene, 2-bromo-1-chloro-4-iodo, acmc-209hpp, 3-bromo-4-chloroiodobenzene, 3-bromo-4-chloro-1-iodobenzene, benzene,2-bromo-1-chloro-4-iodo, 2-bromanyl-1-chloranyl-4-iodanyl-benzene
PubChem CID 50999503
IUPAC Name 2-bromo-1-chloro-4-iodobenzene
SMILES ClC1=C(Br)C=C(I)C=C1

Specifications

Melting Point 39.0°C to 42.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 317.35
Percent Purity 98%
Chemical Name or Material 2-Bromo-1-chloro-4-iodobenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists.

RUO – Research Use Only