missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-phenoxyphenyl)ethanone 95+%, Thermo Scientific™

Catalog No. CC53705DA
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC53705DA 1 g
CC53705DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC53705DA Supplier Thermo Scientific Chemicals Supplier No. CC53705DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28179-33-1
Molecular Formula C14H11BrO2
Molecular Weight (g/mol) 291.14
MDL Number MFCD01419855
InChI Key RAXTYMXDSNWNJS-UHFFFAOYSA-N
Synonym 2-bromo-1-4-phenoxyphenyl ethanone, 2-bromo-1-4-phenoxyphenyl ethan-1-one, ethanone,2-bromo-1-4-phenoxyphenyl, 4-phenoxyphenacyl bromide, 2-bromo-4'-phenoxyacetophenone, 4'-phenoxy-2-bromoacetophenone, alpha-bromo-4'-phenoxyacetophenone, ethanone, 2-bromo-1-4-phenoxyphenyl
PubChem CID 14797675
IUPAC Name 2-bromo-1-(4-phenoxyphenyl)ethanone
SMILES BrCC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 291.14
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(4-phenoxyphenyl)ethanone