missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1,3-thiazol-5-yl]-1-ethanone, ≥90%, Thermo Scientific™

Chemical Identifiers

CAS 423768-43-8
Molecular Formula C10H8BrN3OS
Molecular Weight (g/mol) 298.158
MDL Number MFCD03407321
InChI Key PQWKERRFBZJRSD-UHFFFAOYSA-N
Synonym 2-bromo-1-4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-2-pyrazinyl-5-thiazolyl, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 2-bromo-1 4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethan-1-one
PubChem CID 2776507
IUPAC Name 2-bromo-1-(4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)C2=NC=CN=C2)C(=O)CBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC19805CB
SDS View Documents
Thermo Scientific Chemicals
CC19805CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC19805DA
SDS View Documents
Thermo Scientific Chemicals
CC19805DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC19805DE
SDS View Documents
Thermo Scientific Chemicals
CC19805DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

423768-43-8
298.158
PQWKERRFBZJRSD-UHFFFAOYSA-N
2776507
CC1=C(SC(=N1)C2=NC=CN=C2)C(=O)CBr
C10H8BrN3OS
MFCD03407321
2-bromo-1-4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-2-pyrazinyl-5-thiazolyl, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 2-bromo-1 4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethan-1-one
2-bromo-1-(4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl)ethanone

Specifications

423768-43-8
100.0
Amber Glass Bottle
MFCD03407321
2-bromo-1-4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-2-pyrazinyl-5-thiazolyl, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 2-bromo-1 4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethan-1-one
CC1=C(SC(=N1)C2=NC=CN=C2)C(=O)CBr
298.158
298.16
2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1, 3-thiazol-5-yl]-1-ethanone
89.5
90%
C10H8BrN3OS
250mg
PQWKERRFBZJRSD-UHFFFAOYSA-N
2-bromo-1-(4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl)ethanone
2776507
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents