missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1,3-thiazol-5-yl]-1-ethanone, ≥90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC19805DA

 View more versions of this product

Catalog No. CC19805DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1, 3-thiazol-5-yl]-1-ethanone
89.5
90%
C10H8BrN3OS
1g
PQWKERRFBZJRSD-UHFFFAOYSA-N
2-bromo-1-(4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl)ethanone
2776507
90%
423768-43-8
100.0
Amber Glass Bottle
MFCD03407321
2-bromo-1-4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-2-pyrazinyl-5-thiazolyl, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 2-bromo-1 4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethan-1-one
CC1=C(SC(=N1)C2=NC=CN=C2)C(=O)CBr
298.158
298.16

Chemical Identifiers

423768-43-8
298.158
PQWKERRFBZJRSD-UHFFFAOYSA-N
2776507
CC1=C(SC(=N1)C2=NC=CN=C2)C(=O)CBr
C10H8BrN3OS
MFCD03407321
2-bromo-1-4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-2-pyrazinyl-5-thiazolyl, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 2-bromo-1 4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethan-1-one
2-bromo-1-(4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions