missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-dimethoxybenzene 98.0+%, TCI America™

Catalog No. B34885G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34885G 5 g
B34881G 1 g
B348825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B34885G Supplier TCI America Supplier No. B34885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16932-45-9
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.062
MDL Number MFCD07780177
InChI Key VHVYSMMZHORFKU-UHFFFAOYSA-N
PubChem CID 612296
IUPAC Name 2-bromo-1,3-dimethoxybenzene
SMILES COC1=C(C(=CC=C1)OC)Br

Specifications

Melting Point 95°C
Color White-Yellow
Boiling Point 273°C
Quantity 5 g
Synonym 1-bromo-2,6-dimethoxybenzene, 2-bromo-3-methoxyanisole, 2,6-dimethoxybromobenzene, benzene, bromo-1,3-dimethoxy, benzene,2-bromo-1,3-dimethoxy, 1,3-dimethoxy-2-bromobenzene, acmc-209dzi, 2.6-dimethoxybromobenzene, benzene, 2-bromo-1,3-dimethoxy
Formula Weight 217.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1,3-dimethoxybenzene