missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-dimethoxybenzene, 98%

Catalog No. aab2548506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2548506 5 g
AAB2548503 1 g
AAB2548514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2548506 Supplier Thermo Scientific Chemicals Supplier No. B2548506
Only null left

CAS: 16932-45-9 | C8H9BrO2 | 217.062 g/mol

2-Bromo-1,3-dimethoxybenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,3-dimethoxybenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

Chemical Identifiers

CAS 16932-45-9
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.062
MDL Number MFCD07780177
InChI Key VHVYSMMZHORFKU-UHFFFAOYSA-N
PubChem CID 612296
IUPAC Name 2-bromo-1,3-dimethoxybenzene
SMILES COC1=C(C(=CC=C1)OC)Br

Specifications

Melting Point 91°C to 94°C
Quantity 5 g
Synonym 1-bromo-2,6-dimethoxybenzene, 2-bromo-3-methoxyanisole, 2,6-dimethoxybromobenzene, benzene, bromo-1,3-dimethoxy, benzene,2-bromo-1,3-dimethoxy, 1,3-dimethoxy-2-bromobenzene, acmc-209dzi, 2.6-dimethoxybromobenzene, benzene, 2-bromo-1,3-dimethoxy
Solubility Information Insoluble in water.
Formula Weight 217.06
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,3-dimethoxybenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only