missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-difluorobenzene, 98%

Catalog No. AAA1063314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1063314 25 g
AAA1063306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1063314 Supplier Thermo Scientific Chemicals Supplier No. A1063314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 64248-56-2 | C6H3BrF2 | 192.991 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64248-56-2
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.991
MDL Number MFCD00009894
InChI Key HRZTZLCMURHWFY-UHFFFAOYSA-N
Synonym 1-bromo-2,6-difluorobenzene, 2,6-difluorobromobenzene, benzene, 2-bromo-1,3-difluoro, 2-bromo-1,3-difluoro-benzene, m-difluorobromobenzene, pubchem3034, acmc-1bc5o, 2,6-difluoro-bromobenzene, 1-bromo-2,6-diflurobenzene
PubChem CID 123557
IUPAC Name 2-bromo-1,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)Br)F

Specifications

Density 1.71
Boiling Point 61°C (35 mmHg)
Flash Point 53°C (127°F)
Refractive Index 1.511
Quantity 25 g
UN Number UN1993
Beilstein 1681053
Formula Weight 192.99
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,3-difluorobenzene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 264-750-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only