missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-2-tetralone, 95%, Thermo Scientific™

Catalog No. AC294970050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC294970050 Supplier Thermo Scientific Chemicals Supplier No. 294970050
Only null left

Chemical Identifiers

CAS 4133-34-0
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00001730
InChI Key XEAPZXNZOJGVCZ-UHFFFAOYSA-N
Synonym 7-methoxy-2-tetralone, 7-methoxy-3,4-dihydronaphthalen-2 1h-one, 3,4-dihydro-7-methoxy-2 1h-naphthalenone, 7-methoxy-3,4-dihydronaphthalen-2-one, 7-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 7-methoxyl-2-tetralone, 7-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxy-1,3,4-trihydronaphthalen-2-one, 3,4-dihydro-7-methoxynaphthalen-2 1h-one
PubChem CID 77785
IUPAC Name 7-methoxy-3,4-dihydro-1H-naphthalen-2-one
SMILES COC1=CC2=C(CCC(=O)C2)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Orange
Density 1.1300g/mL
Boiling Point 124°C to 126°C (1.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5590 to 1.563
Quantity 5 g
Specific Gravity 1.13
Formula Weight 176.22
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 7-Methoxy-2-tetralone
Show More Show Less
EINECSNumber 223-954-1

RUO – Research Use Only