missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,5-trimethyl-4-nitrobenzene, 90+%

Catalog No. AAL0510703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0510703 1 g
AAL0510706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0510703 Supplier Thermo Scientific Chemicals Supplier No. L0510703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 90561-85-6 | C9H10BrNO2 | 244.088 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90561-85-6
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD00015924
InChI Key IUFDRRDWXUSFBG-UHFFFAOYSA-N
Synonym 3-bromo-2,4,6-trimethylnitrobenzene, 2-bromo-4-nitro-1,3,5-trimethylbenzene, 1-bromo-3-nitro-2,4,6-trimethylbenzene, 4-bromo-2-nitromesitylene, 4-bromo-1,3,5-trimethyl-2-nitrobenzene, 2-bromanyl-1,3,5-trimethyl-4-nitro-benzene
PubChem CID 2757200
IUPAC Name 2-bromo-1,3,5-trimethyl-4-nitrobenzene
SMILES CC1=CC(=C(C(=C1[N+](=O)[O-])C)Br)C

Specifications

Melting Point 56°C to 62°C
Boiling Point 106°C to 107°C (1.3 mmHg)
Quantity 1 g
Beilstein 1958703
Sensitivity Light sensitive
Formula Weight 244.09
Percent Purity ≥90%
Chemical Name or Material 2-Bromo-1,3,5-trimethyl-4-nitrobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only