missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,5-triisopropylbenzene 95.0+%, TCI America™

Catalog No. p-7122995
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35135G 5 g
B351325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35135G Supplier TCI America Supplier No. B35135G
Only null left

Chemical Identifiers

CAS 21524-34-5
Molecular Formula C15H23Br
Molecular Weight (g/mol) 283.253
MDL Number MFCD00051547
InChI Key FUMMYHVKFAHQST-UHFFFAOYSA-N
Synonym 2-bromo-1,3,5-triisopropylbenzene, 1-bromo-2,4,6-triisopropylbenzene, 2,4,6-triisopropylbromobenzene, 1,3,5-triisopropyl-2-bromobenzene, 2-bromo-1,3,5-tris propan-2-yl benzene, 2-bromo-1,3,5-triisopropyl-benzene, 2-bromo-1,3,5-tri propan-2-yl benzene, benzene, 2-bromo-1,3,5-tris 1-methylethyl, 2-bromo-1,3,5-tris 1-methylethyl-benzene, acmc-209flf
PubChem CID 140846
IUPAC Name 2-bromo-1,3,5-tri(propan-2-yl)benzene
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)Br)C(C)C

Specifications

Color Yellow
Boiling Point 148°C
Quantity 5 g
Formula Weight 283.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-1,3,5-triisopropylbenzene