missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2-thienyl)-1-ethanone, Technical Grade, Thermo Scientific™

Chemical Identifiers

CAS 10531-41-6
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.069
InChI Key UHWNENCHFSDZQP-UHFFFAOYSA-N
Synonym 2-bromo-1-thiophen-2-yl ethanone, 2-2-bromoacetyl thiophene, 2-bromo-1-2-thienyl-1-ethanone, 2-bromoacetyl thiophene, 2-bromo-1-thiophen-2-yl-ethanone, 2-bromo-1-2-thienyl ethanone, 2-bromo-1-thiophen-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thienyl, ethanone, 2-bromo-1-thienyl, 2-bromo-1-2-thienyl ethan-1-one
PubChem CID 2776372
IUPAC Name 2-bromo-1-thiophen-2-ylethanone
SMILES C1=CSC(=C1)C(=O)CBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC13005CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10531-41-6
205.069
2-bromo-1-thiophen-2-yl ethanone, 2-2-bromoacetyl thiophene, 2-bromo-1-2-thienyl-1-ethanone, 2-bromoacetyl thiophene, 2-bromo-1-thiophen-2-yl-ethanone, 2-bromo-1-2-thienyl ethanone, 2-bromo-1-thiophen-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thienyl, ethanone, 2-bromo-1-thienyl, 2-bromo-1-2-thienyl ethan-1-one
2-bromo-1-thiophen-2-ylethanone
C6H5BrOS
UHWNENCHFSDZQP-UHFFFAOYSA-N
2776372
C1=CSC(=C1)C(=O)CBr

Specifications

10531-41-6
C6H5BrOS
UHWNENCHFSDZQP-UHFFFAOYSA-N
2-bromo-1-thiophen-2-ylethanone
2776372
90%
90%
2-bromo-1-thiophen-2-yl ethanone, 2-2-bromoacetyl thiophene, 2-bromo-1-2-thienyl-1-ethanone, 2-bromoacetyl thiophene, 2-bromo-1-thiophen-2-yl-ethanone, 2-bromo-1-2-thienyl ethanone, 2-bromo-1-thiophen-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thienyl, ethanone, 2-bromo-1-thienyl, 2-bromo-1-2-thienyl ethan-1-one
C1=CSC(=C1)C(=O)CBr
205.069
205.07
2-Bromo-1-(2-thienyl)-1-ethanone
Videos
SDS
Documents

Documents